missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 5-chloromethyl-1,3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dimethylpyrazole, 1h-pyrazole, 5-chloromethyl-1,3-dimethyl, 5-chloromethyl-1, pubchem23625, 3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dime..., 5-chloromethyl-1,4-dimethyl-1h-pyrazole, 1h-pyrazole,5-chloromethyl-1,3-dimethyl |
IUPAC Name | 5-(chloromethyl)-1,3-dimethylpyrazole |
Formula Weight | 144.6 |
Percent Purity | 97% |
Chemical Name or Material | 5-(Chloromethyl)-1, 3-dimethyl-1H-pyrazole |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction