missing translation for 'onlineSavingsMsg'
Learn More
5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Thermo Scientific™ Product Code.: 10182924

5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Thermo Scientific™

Product Code. 10182924
5 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10182924

Brand: Thermo Scientific Maybridge CC12524DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852227-86-2
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD07368502
InChI Key SGEZKPNUNBVVLB-UHFFFAOYSA-N
PubChem CID 4961270
SMILES CN1N=C(C)C=C1CCl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 5-chloromethyl-1,3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dimethylpyrazole, 1h-pyrazole, 5-chloromethyl-1,3-dimethyl, 5-chloromethyl-1, pubchem23625, 3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dime..., 5-chloromethyl-1,4-dimethyl-1h-pyrazole, 1h-pyrazole,5-chloromethyl-1,3-dimethyl
IUPAC Name 5-(chloromethyl)-1,3-dimethylpyrazole
Formula Weight 144.6
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-1, 3-dimethyl-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.