missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Chloromethyl)-3-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 200.65 |
Percent Purity | 95% |
Chemical Name or Material | 5-(Chloromethyl)-3-(2-thienyl)-1, 2, 4-oxadiazole |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-(Chloromethyl)-3-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction