missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals (+)-5-Fluoro-2'-deoxyuridine, 99+% Product Code.: 10124860

Thermo Scientific Chemicals (+)-5-Fluoro-2'-deoxyuridine, 99+%

Product Code. 10124860
500 mg, Glass bottle
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10124860

Brand: Thermo Scientific Chemicals 227605000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-91-9
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00006530
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurdShow MoreShow Less
PubChem CID 5790
ChEBI CHEBI:60761
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 148.0°C to 153.0°C
Color White
Loss on Drying 0.5% (60°C, 4 hrs) (vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Merck Index 15,4144
Specific Rotation Condition + 35.90 (20.00°C c=1,water)
Specific Rotation + 35.90
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,practically insoluble in chcl3,insoluble in ether and hexane
Formula Weight 246.19
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material (+)-5-Fluoro-2′-deoxyuridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464240 Certificate TypeCertificate of Analysis Date12/07/2024 Catalog Number227601000,227600025,227605000
Lot NumberA0456661 Certificate TypeCertificate of Analysis Date28/09/2023 Catalog Number227601000,227600025,227605000
Lot NumberA0398357 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number227601000,227600025,227605000
Lot NumberA0409608 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number227601000,227600025,227605000
Lot NumberA0406019 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number227601000,227600025,227605000
compliance-icons compliance-icons
Product Identifier
  • (+)-5-Fluoro-2'-deoxyuridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals (+)-5-Fluoro-2'-deoxyuridine, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.