missing translation for 'onlineSavingsMsg'
Learn More
5-Fluorobenzoxazole, 97%, Thermo Scientific™ Product Code.: 15355028

5-Fluorobenzoxazole, 97%, Thermo Scientific™

Product Code. 15355028
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15355028

Brand: Thermo Scientific Acros 455612500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 221347-71-3
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.11
InChI Key ZRMPAEOUOPNNPZ-UHFFFAOYSA-N
Synonym 5-fluorobenzoxazole, 5-fluorobenzo d oxazole, benzoxazole, 5-fluoro, benzoxazole,5-fluoro, pubchem21888, 5-fluoranyl-1,3-benzoxazole, benzoxazole, 5-fluoro-9ciShow MoreShow Less
PubChem CID 19937167
IUPAC Name 5-fluoro-1,3-benzoxazole
SMILES C1=CC2=C(C=C1F)N=CO2

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 250 mg
Formula Weight 137.11
Percent Purity 97%
Chemical Name or Material 5-Fluorobenzoxazole
compliance-icons
Product Identifier
  • 5-Fluorobenzoxazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Fluorobenzoxazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.