missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-4H-chromen-4-one, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 162.14 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 5-Hydroxy-4H-chromen-4-one |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Hydroxy-4H-chromen-4-one, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction