missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

75.70 € - 610.00 €

Chemical Identifiers

CAS 98700-50-6
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
InChI Key IIEFFVFWBUGUTI-UHFFFAOYSA-N
PubChem CID 2776437
IUPAC Name 5-methyl-1-phenylpyrazole-4-carbaldehyde
SMILES CC1=C(C=NN1C2=CC=CC=C2)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10282582
View Documents
Thermo Scientific Maybridge
CC15504DA
1 g Amber Glass Bottle
75.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
11360577
View Documents
Thermo Scientific Maybridge
CC15504EA
10 g Amber Glass Bottle
610.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

98700-50-6
186.214
2776437
CC1=C(C=NN1C2=CC=CC=C2)C=O
C11H10N2O
IIEFFVFWBUGUTI-UHFFFAOYSA-N
5-methyl-1-phenylpyrazole-4-carbaldehyde

Specifications

98700-50-6
C11H10N2O
IIEFFVFWBUGUTI-UHFFFAOYSA-N
5-methyl-1-phenylpyrazole-4-carbaldehyde
2776437
97%
97%
5-methyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 5-methyl-1-phenyl-1h-pyrazole-4-carboxaldehyde, acmc-20am2d, 1h-pyrazole-4-carboxaldehyde, 5-methyl-1-phenyl, 1h-pyrazole-4-carboxaldehyde,5-methyl-1-phenyl
CC1=C(C=NN1C2=CC=CC=C2)C=O
186.214
186.21
5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.