missing translation for 'onlineSavingsMsg'
Learn More
5-Methylthiazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 15489558

5-Methylthiazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15489558
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15489558

Brand: Thermo Scientific Alfa Aesar H63076.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5-Methylthiazole-2-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylthiazole-2-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61291-21-2
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD08060736
InChI Key UYDGNRLMYZMDOG-UHFFFAOYSA-N
Synonym 5-methylthiazole-2-carboxylic acid, 5-methyl-thiazole-2-carboxylic acid, 5-methyl-2-thiazolecarboxylic acid, 2-thiazolecarboxylic acid, 5-methyl, 2-carboxy-5-methyl-1,3-thiazole, pubchem23003, acmc-209mr5, ksc352m3t, 5-methyl thiazole-2-carboxylic acidShow MoreShow Less
PubChem CID 21877349
IUPAC Name 5-methyl-1,3-thiazole-2-carboxylic acid
SMILES CC1=CN=C(S1)C(O)=O

Specifications

Color White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 143.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Methylthiazole-2-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5006R29T Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63076.03,H63076.MD,H63076.06
Lot Number81400436 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63076.03,H63076.MD,H63076.06
Lot NumberX06B006 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63076.03,H63076.MD,H63076.06
Lot Number5038F28P Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63076.03,H63076.MD,H63076.06
Lot Number81301160 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63076.03,H63076.MD,H63076.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Methylthiazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.