Learn More
Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
For smaller dialysis experiments or evaluating dialysis tubing for dialysis application
Brand:  Spectrum™ 132 703T1
 View more versions of this product
Product Code. 15300772
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes:
  • 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
  • One Standard Spectra/Por Closure (for buoyance)
  • One weighted Spectra/Por Closure (for orientation)
  • Two adhesive tubing openers
Description
Description
  • Hydrophilic, symmetric porosity
  • Ensures higher permeability and basic dialysis
  • Available as dry with glycerin (soak in water)
Specifications
Specifications
12 to 14 kDa
20.4 mm
3.3 mL/cm vol./length
1 m Tubing/Kit