missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
For smaller dialysis experiments or evaluating dialysis tubing for dialysis application
47.85 € - 209.00 €
Specifications
MWCO | 12 to 14 kDa |
---|
Includes:
- 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
- One Standard Spectra/Por Closure (for buoyance)
- One weighted Spectra/Por Closure (for orientation)
- Two adhesive tubing openers
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
15320772
|
Spectrum™
132709T |
18 mL/cm vol./length | 48 mm | 5 m Tubing/Kit |
209.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15330772
|
Spectrum™
132 709T1 |
18 mL/cm vol./length | 48 mm | 1 m Tubing/Kit | |||||
15300772
|
Spectrum™
132 703T1 |
3.3 mL/cm vol./length | 20.4 mm | 1 m Tubing/Kit | |||||
15390762
|
Spectrum™
132703T |
3.3 mL/cm vol./length | 20.4 mm | 5 m Tubing/Kit |
189.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Hydrophilic, symmetric porosity
- Ensures higher permeability and basic dialysis
- Available as dry with glycerin (soak in water)
Specifications
12 to 14 kDa |