missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs 0.5 - 1.0 kD Biotech CE Dialysis Kits

With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane

Brand:  Spectrum™ 131090T

174.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11550940

  • 179.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:
  • 1m roll of Biotech CE Dialysis Tubing (16mm flatwidth)
  • 2 Universal tubing closures
  • 5 Tubing opening picks
This item is not returnable. View return policy

Description

Description

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks

Specifications

Specifications

Dialysis Trial Kit
Cellulose Ester
0.39 in.
1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks
Ideal for small, critical dialysis applications
0.5 to 1 kDa
10 mm
0.79 mL/cm vol./length
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.