Learn More
Spectrum™ Labs 0.5 - 1.0 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
166.00 € - 308.00 €
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
15370662
|
Spectrum™
131096T |
20 mm | 3.1 mL/cm vol./length |
308.00 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15360662
|
Spectrum™
131093T |
15 mm | 1.8 mL/cm vol./length |
203.00 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15350662
|
Spectrum™
131084T |
6.4 mm | 0.32 mL/cm vol./length |
166.00 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11550940
|
Spectrum™
131090T |
10 mm | 0.79 mL/cm vol./length |
179.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks