missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor for Centrifuge 5810 / 5810 R
Eppendorf Rotor for Centrifuge 5810/5810 R - precision redefined. High-capacity centrifugation with varied rotor options, a range of capacities and speeds, and versatile buckets to ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
Brand: Eppendorf™ 5820740000
3460.19 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.

Specifications
S-4-104 - Includes 750 mL round buckets (x4) | |
4 x 750 mL | |
3,900 rpm | |
750 mL Round Buckets (x4) |
Rotor | |
3,214 x g | |
Centrifuge 5810 / 5810 R |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ Rotor for Centrifuge 5810 / 5810 R > Description: S-4-104 - Includes 750 mL round buckets (x4), Capacity: 4 x 750 mL, Max. RCF: 3,214 x g, Max. Speed: 3,900 rpm
Spot an opportunity for improvement?Share a Content Correction