Learn More
5X Kinase Buffer A
5X Kinase Buffer A
Brand:  Invitrogen PV6135
 View more versions of this product
Product Code. 11300972
  • 742.00 € / Each
Explore more special offers
Description
Description
The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.
Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays
  • Optimized 5X concentrated formulation
  • Simplified handling with room temperature storage
    Easily Screen for Kinase Targets using an Invitrogen™ Platform
    For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:
    LanthaScreen™ Eu Binding Assay
    LanthaScreen™ Activity Assay
    Adapta™ Universal Kinase Assay
    Z´-LYTE™ Kinase Assay Kits
    Save Freezer Space
    The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.
    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    5X
    Pharma & Biopharma; Protein Assays and Analysis
    250 mM HEPES (pH 7.5), 50 mM MgCl2, 5 mM EGTA, and 0.05% Brij-35
    100 ml
    Liquid
    7.5
    Adapta™ and Z-LYTE™ assays;
    Approved for shipment at Room Temperature or on Wet or Dry Ice
    Kinase Buffer A