Learn More
5X Kinase Buffer A
5X Kinase Buffer A
Brand:  Invitrogen PV3189
 View more versions of this product
Product Code. 10013013
  • 200.00 € / Each
Explore more special offers
Description
Description
The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.
Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays
  • Optimized 5X concentrated formulation
  • Simplified handling with room temperature storage
    Easily Screen for Kinase Targets using an Invitrogen™ Platform
    For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:
    LanthaScreen™ Eu Binding Assay
    LanthaScreen™ Activity Assay
    Adapta™ Universal Kinase Assay
    Z´-LYTE™ Kinase Assay Kits
    Save Freezer Space
    The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.
    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    5X
    5X Kinase Buffer A may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
    Approved for shipment at Room Temperature or on Wet or Dry Ice
    4 mL
    Liquid
    Pharma & Biopharma; Protein Assays and Analysis
    Biochemical binding/kinetic, Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity
    ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA T674I, PDGFRA V561D, PTK6, RET Y791F, SYK, TYRO3, ZAP70, MAP4K5, MST4, PRKCQ, PRKX, ROCK2, IKBKE, PAK4, EIF2AK1, MAP3K2, LRRK2 D1994A, GRK7, GSK3A, GSK3B, SNF1LK2, ACVR2A, BMPR1A, MAP2K1 S218D S222D, RAF1 (cRAF) Y340D Y341D, FGFR1, FGFR2, PRKCD, MAPK13, MAPK8, MAP3K10, MAP3K9, JAK1, JAK2 JH1 JH2, AMPK (A2/B1/G1), CAMK1, CAMK2B, CHEK1, MAPKAPK2, PASK, PIM2, PRKCN, PRKD2, PRKG2, RPS6KA5, MELK, STK33, BLK, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, ROS1, TEK, TTK, NEK2, PRKCG, SGK, CHUK, IKBKB, TAOK3, JAK2 JH1 JH2 V617F, TYK2, DNAPK, IRAK1, ACVR1B, MAPKAPK5, MYLK2, SRPK2, STK4, WNK2, FLT1, MUSK, SRCN1, TEC, CAMK2D, MAP3K7/MAP3K7IP1, LRRK2 R1441C, CSNK1G1, DAPK2, DYRK1A, DYRK3, HIPK3, MAP2K1, MAP2K6, PIK3CG, RPS6KA6, DDR1, RIPK2, ABL1 E255K, BMX, EPHA4, FYN, PTK2, PTK2B, RET, MAP4K2, MINK1, SGK2, GRK6, MAPK9, JAK3, LRRK2, AMPK (A1/B1/G1), AURKA, CAMK1D, CAMK4, CHEK2, MAPKAPK3, PIM1, PKN1, PRKD1, PRKG1, RPS6KA4, STK22D, CSNK1G2, NUAK1, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, PRKCA, PRKCI, CAMK2A, PAK1, RPS6KA3, MAPK14, EIF2AK3, MAP3K3, WNK3, ABL1, ABL2, AXL, FGFR3, FGFR3 K650E, FGR, HCK, KIT V654A, AURKB, TAOK2, MAP3K11, CAMKK1, CDC42BPA, CDK1, CDK5, CDK8, CDK9/Cyclin K, MAP2K3, TGFBR1, NLK, MAPK10, CSNK1G3, DMPK, DYRK1B, ACVR1, TGFBR2, EPHA1, EPHA8, EPHB3, INSRR, LYNB, MST1R, NTRK1, PDGFRA D842V, SRMS, MAP4K4, AKT2, LRRK2 G2019S, CAMKK2, CDC42BPB, CDK2, CDK7, CDK9/Cyclin T1, CSNK1E, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, PDGFRB, MARK4, RPS6KB1, EGFR T790M, EPHA7, EPHB1, MET M1250T, DYRK4, PHKG1, PRKCZ, ROCK1, RPS6KA1, TBK1, MAPK1, MAPK11, BRAF V599E, JAK2, MARK1, PHKG2, MLCK, PAK6, SLK, LIMK1, MKNK2, MYLK, FRK, ITK, PRKCB1, STK24, AKT3, BRAF, LRRK2 I2020T, BRSK1, CLK2, PDK1, MAP2K2, PIK3CA/PIK3R1, PIK3CD/PIK3R1, ACVR2B, MAP3K14, MAP2K6 S207E T211E, FLT3 D835Y, SRC, GRK5, GRK4, STK3, ABL1 Y253F, BTK, DDR2, FES, KIT T670I, TXK, PRKCB2, RPS6KA2, SGKL, STK25, AKT1, PAK7, PRKACA, MAPK12, MAPK3, MAP3K8, CLK1, CLK4, MARK2, STK17A, ERBB2, NTRK2, PRKCE, PRKCH, EIF2AK2, MAP3K5, IRAK4, EGFR T790M L858R, LIMK2, STK16
    Kinase Buffer A