missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-chloroimidazo[2,1-b][1,3]thiazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | C6H5CH2CH2NCS |
Quantity | 5 g |
Synonym | 6-chloroimidazo 2,1-b thiazole, 6-chloroimidazo 2,1-b 1,3 thiazole, 6-chloroimidazole 2,1-b 1,3 thiazole, 6-chloroimidazo 2,1-b 1,3-thiazoline, chloroimidazobthiazole, acmc-1cip3, 6-chloro-imidazo 2,1-b thiazole, imidazo 2,1-b thiazole,6-chloro, imidazo 2,1-b thiazole, 6-chloro, 6-chloro-4h-imidazo 2,1-b thiazole |
Formula Weight | 158.61 |
Percent Purity | 97% |
Chemical Name or Material | 6-chloroimidazo[2, 1-b][1, 3]thiazole |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
6-chloroimidazo[2,1-b][1,3]thiazole, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction