missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Thermo Scientific™

30.60 € - 378.00 €

Chemical Identifiers

CAS 6272-26-0
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00068174
InChI Key GBDMODVZBPFQKI-UHFFFAOYSA-N
Synonym 6-hydroxybenzofuran-3 2h-one, 6-hydroxy-2h-benzofuran-3-one, 6-hydroxy-2,3-dihydrobenzo b furan-3-one, 6-hydroxy-1-benzofuran-3 2h-one, 6-hydroxybenzofuran-3-one, 3 2h-benzofuranone, 6-hydroxy, 6-hydroxy-3-coumaranone, 6-hydroxy-2,3-dihydro-1-benzofuran-3-one, 6-hydroxy-3 2h-benzofuranone, 6-hydroxy-2h-1-benzofuran-3-one
PubChem CID 235717
IUPAC Name 6-hydroxy-1-benzofuran-3-one
SMILES OC1=CC=C2C(=O)COC2=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10273372
View Documents Promotion Details
Thermo Scientific Maybridge
SEW02738DA
1 g Amber Glass Bottle
30.60 €
1g
Please to purchase this item. Need a web account? Register with us today!
10233422
View Documents Promotion Details
Thermo Scientific Maybridge
SEW02738EA
10 g Amber Glass Bottle
174.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10747685
View Documents Promotion Details
Thermo Scientific Maybridge
SEW02738EB
25 g Amber Glass Bottle
378.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6272-26-0
150.13
GBDMODVZBPFQKI-UHFFFAOYSA-N
235717
OC1=CC=C2C(=O)COC2=C1
C8H6O3
MFCD00068174
6-hydroxybenzofuran-3 2h-one, 6-hydroxy-2h-benzofuran-3-one, 6-hydroxy-2,3-dihydrobenzo b furan-3-one, 6-hydroxy-1-benzofuran-3 2h-one, 6-hydroxybenzofuran-3-one, 3 2h-benzofuranone, 6-hydroxy, 6-hydroxy-3-coumaranone, 6-hydroxy-2,3-dihydro-1-benzofuran-3-one, 6-hydroxy-3 2h-benzofuranone, 6-hydroxy-2h-1-benzofuran-3-one
6-hydroxy-1-benzofuran-3-one

Specifications

6272-26-0
97%
MFCD00068174
GBDMODVZBPFQKI-UHFFFAOYSA-N
6-hydroxy-1-benzofuran-3-one
235717
97%
97.0
C8H6O3
6-hydroxybenzofuran-3 2h-one, 6-hydroxy-2h-benzofuran-3-one, 6-hydroxy-2,3-dihydrobenzo b furan-3-one, 6-hydroxy-1-benzofuran-3 2h-one, 6-hydroxybenzofuran-3-one, 3 2h-benzofuranone, 6-hydroxy, 6-hydroxy-3-coumaranone, 6-hydroxy-2,3-dihydro-1-benzofuran-3-one, 6-hydroxy-3 2h-benzofuranone, 6-hydroxy-2h-1-benzofuran-3-one
OC1=CC=C2C(=O)COC2=C1
150.13
150.13
6-Hydroxy-2, 3-dihydrobenzo[b]furan-3-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications