missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ 6-IAF (6-Iodoacetamidofluorescein)

6-IAF (6-Iodoacetamidofluorescein)

Brand:  Molecular Probes™ I30452

554.80 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 11564896

  • 600.00 € / 25mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The thiol-reactive 6-iodoacetamidofluorescein (6-IAF) can be used to produce bioconjugates with the 5-isomer of fluorescein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
FITC (Fluorescein)
6-IAF
Thiol
Alkyl Halide, Iodoacetamide
Classic Dyes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2725290 Certificate TypeCertificate of Analysis Date03/08/2023 Catalog NumberI30452
Lot Number2486536 Certificate TypeCertificate of Analysis Date06/02/2023 Catalog NumberI30452
Lot Number2415849 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog NumberI30452
Lot Number2415848 Certificate TypeCertificate of Analysis Date16/09/2021 Catalog NumberI30452
Lot Number2295073 Certificate TypeCertificate of Analysis Date15/07/2021 Catalog NumberI30452
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ 6-IAF (6-Iodoacetamidofluorescein) > 25mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.