missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
Specifications
Assay Percent Range | 90% |
Packaging | Amber glass bottle |
Quantity | 50 g |
IUPAC Name | 3-methyl-1H-pyridazin-6-one;hydrate |
Formula Weight | 128.14 |
Percent Purity | 90% |
Chemical Name or Material | 6-Methyl-2, 3-dihydropyridazin-3-one hydrate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction