missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 158.12 |
Percent Purity | 97% |
Chemical Name or Material | 6-Hydroxy-3-pyridazinecarboxylic acid monohydrate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction