Learn More
YSI™ 6051010 Pro Series ISE/ISE Cable
Measure any combination of ISEs (pH, ORP, ammonium, nitrate, chloride) and temperature
Brand:  YSI™ 6051010-10
 View more versions of this product
Product Code. 12903497
  • 815.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The 6051010 cables ONLY work with the YSI Pro Plus handheld
Order Info
Order probes separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cable management kit
393.7 in.
10 m
ProPlus handheld instrument