Learn More
YSI™ 6051010 Pro Series ISE/ISE Cable
Measure any combination of ISEs (pH, ORP, ammonium, nitrate, chloride) and temperature
744.00 € - 1095.00 €
Product Code | Brand | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
12923497
|
YSI™
6051010-30 |
1181.1 in. | 30 m |
1095.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
12993487
|
YSI™
6051010-4 |
157.4 in. | 4 m |
744.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
12903497
|
YSI™
6051010-10 |
393.7 in. | 10 m |
815.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
12913497
|
YSI™
6051010-20 |
787.4 in. | 20 m |
954.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
Description
The 6051010 cables ONLY work with the YSI Pro Plus handheld
Order Info
Order probes separately.