Learn More
YSI™ 6051010 Pro Series ISE/ISE Cable
Measure any combination of ISEs (pH, ORP, ammonium, nitrate, chloride) and temperature
Brand:  YSI™ 6051010-20
 View more versions of this product
Product Code. 12913497
  • 954.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The 6051010 cables ONLY work with the YSI Pro Plus handheld
Order Info
Order probes separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cable management kit
787.4 in.
20 m
ProPlus handheld instrument