missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ 70-205 Series Yellow Neptune™ Light Weight Gloves

Designed to provide mechanical strength in light-to-heavy duty handling and production applications. Neptune™ Aramid fiber Industrial Knitted Gloves offer enhanced comfort and dexterity.

119.00 €

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003,EN 407:2004
Classification Standards CATEGORY III EN 388 : 134X, EN 407, ANSI
Description Knit construction
Color Yellow
Cut Resistance Rating Very high cut resistance
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12690962
View Documents Promotion Details
Ansell™
70205100
10
119.00 €
12 pairs
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
12610972
View Documents Promotion Details
Ansell™
70205080
8
119.00 €
12 pairs
In Stock
to see stock.
Edge
Add to basket
 
12600972
View Documents Promotion Details
Ansell™
70205070
7
119.00 €
12 pairs
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
12620972
View Documents Promotion Details
Ansell™
70205090
9
119.00 €
12 pairs
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
Description

Description

  • High-cut resistance with Kevlar™ aramid fibers reinforcement
  • Neptune coating allows glove fibers to repel liquids
  • Reinforced thumb welt increases abrasion resistance
  • Ambidextrous
  • Washable and reusable
  • Knitwrist cuff
Specifications

Specifications

EN 420:2003 + A1:2009 and EN 388:2003,EN 407:2004
Knit construction
Very high cut resistance
13 gauge
12 pairs in a bag, 12 bags in a carton
70-205
Standards CATEGORY III EN 388 : 134X, EN 407, ANSI
Yellow
Mechanical Protection Gloves
Knit
144 Pairs
220 to 280 mm
Videos
SDS
Documents

Documents

Product Certifications