Learn More
Hamilton™ 700 Microliter Syringes: N Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
67.15 € - 149.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
For Use With (Application) | For dispensing low volumes to achieve the highest level of accuracy and precision possible |
Length (English) Needle | 2 in. |
Product Type | Microliter Syringe |
Product Code | Brand | Volume (Metric) | Needle Point Style | Diameter (Metric) Inner Needle | Diameter (Metric) Outer Needle | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Point Style | Diameter (Metric) Inner Needle | Diameter (Metric) Outer Needle | Price | Quantity & Availability | ||
10058300
|
Hamilton™
80765 |
250 μL | Blunt | 0.41 mm | 0.72 mm |
128.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10095630
|
Hamilton™
80865 |
500 μL | Blunt | 0.41 mm | 0.72 mm |
114.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10054414
|
Hamilton™
80839 |
500 μL | Cone | 0.41 mm | 0.72 mm |
149.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10097671
|
Hamilton™
80339 |
10 μL | Cone | 0.13 mm | 0.47 mm |
96.50 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10313811
|
Hamilton™
80383 |
10 μL | Blunt | 0.13 mm | 0.72 mm |
121.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10180361
|
Hamilton™
80565 |
50 μL | Blunt | 0.15 mm | 0.72 mm |
115.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10778541
|
Hamilton™
80465 |
25 μL | Blunt | 0.15 mm | 0.72 mm |
117.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10340711
|
Hamilton™
80365 |
10 μL | 3 | 0.15 mm | 0.72 mm |
67.15 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10261821
|
Hamilton™
80500 |
50 μL | Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm |
99.20 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10429042
|
Hamilton™
80384 |
10 μL | Sharp, Bevel, Curved, Non-coring | 0.13 mm | 0.47 mm |
94.65 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10149980
|
Hamilton™
80400 |
25 μL | Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm |
84.65 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10020332
|
Hamilton™
87919 |
5 μL | 3 | 0.13 mm | 0.483 mm |
80.05 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10313571
|
Hamilton™
80600 |
100 μL | Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm |
147.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10625251
|
Hamilton™
80665 |
100 μL | Blunt | 0.15 mm | 0.72 mm |
133.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10251871
|
Hamilton™
80700 |
250 μL | Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10303621
|
Hamilton™
80800 |
500 μL | Sharp, Bevel, Curved, Non-coring | 0.41 mm | 0.72 mm |
133.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Original hand-fitted Hamilton syringe
- Ideal for dispensing volumes from 0.5 µL up to 500 µL
- A variety of different terminations and needle options are available in this series
Specifications
Not Autoclavable | |
For dispensing low volumes to achieve the highest level of accuracy and precision possible | |
2 in. | |
Microliter Syringe |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hamilton™ 700 Microliter Syringes: N Termination