Learn More
Hamilton™ 700 Series Cemented Needle Syringes
This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
Brand:  Hamilton™ DS80465
 View more versions of this product
Product Code. 10569881
  • 2010.00 € / Each
Explore more special offers
Description
Description
  • The original hand-fitted Hamilton™ syringe
  • Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
  • Unsurpassed syringe life
Specifications
Specifications
Borosilicate Glass, Stainless Steel
7.75 mm
1/Ea.
3
22 G
Syringe
25 μL
Cemented