Learn More
Hamilton™ 700 Series Cemented Needle Syringes
This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
68.15 € - 2010.00 €
Specifications
Material | Borosilicate Glass, Stainless Steel |
---|---|
Connection Type | Cemented |
Product Code | Brand | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
10569881
|
Hamilton™
DS80465 |
25 μL | 22 G |
2010.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11528034
|
Hamilton™
DS80865 |
500 μL | 22 G |
2010.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10680812
|
Hamilton™
80350 |
10 μL | 26s G |
68.15 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The original hand-fitted Hamilton™ syringe
- Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
- Unsurpassed syringe life
Specifications
Borosilicate Glass, Stainless Steel | |
Cemented |