missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Q705 Sonicator without Probe

Guarantees more efficient operation, sample-to-sample consistency and most importantly, a reliable end result. Fisherbrand™ Q705 Sonicator without Probe is the most technologically advanced sonicator available today.

Brand:  Fisherbrand™ FB705AEUK-220

Product Code. 12971151

  • 4990.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Full amplitude control from 1-100%
  • Programmability including processing times, pulse on/off and memory
  • Optional temperature monitor probe
  • Built lead-free
  • Multiple programs can be run in sequence
  • Total energy output display
  • Overload protection
Specifications

Specifications

110 V
700 W
Generator, converter, power cable, converter set, wrench set
20 kHz
387 x 203 x 216 mm (15.25 x 8 x 8.5 in.)
Q705
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Q705 Sonicator without Probe > Capacity: 0.2 to 1000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.