missing translation for 'onlineSavingsMsg'
Learn More
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals Product Code.: 10274730

8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals

Product Code. 10274730
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10274730

Brand: Thermo Scientific Acros 121901000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-24-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006807
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
PubChem CID 1923
ChEBI CHEBI:48981
IUPAC Name quinolin-8-ol
SMILES OC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 72.0°C to 74.0°C
Color Beige or White to Yellow
Boiling Point 267.0°C (752.0 mmHg)
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Beilstein 21, IV, 1135
Merck Index 15, 4881
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,chloroform and,benzene
Formula Weight 145.16
Grade ACS Reagent
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 8-Hydroxyquinoline, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458633 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number121900250,121900010,121901000,121905000,121900025
Lot NumberA0450707 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number121900250,121900010,121901000,121905000,121900025
Lot NumberA0409041 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number121900250,121900010,121901000,121905000,121900025
Lot NumberA0378412 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number121900250,121900010,121901000,121905000,121900025
Lot NumberA0444363 Certificate TypeCertificate of Analysis Date20/07/2022 Catalog Number121900250,121900010,121901000,121905000,121900025
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.