missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator

Specifications
Specifications
Melting Point | 72.0°C to 74.0°C |
Color | Beige or White to Yellow |
Boiling Point | 267.0°C (752.0 mmHg) |
Ignition Residue | 0.05% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 25 g |
Beilstein | 21, IV, 1135 |
Merck Index | 15, 4881 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458633 | Certificate of Analysis | 11/12/2023 | 121900250,121900010,121901000,121905000,121900025 |
A0450707 | Certificate of Analysis | 02/05/2023 | 121900250,121900010,121901000,121905000,121900025 |
A0409041 | Certificate of Analysis | 13/12/2022 | 121900250,121900010,121901000,121905000,121900025 |
A0378412 | Certificate of Analysis | 13/12/2022 | 121900250,121900010,121901000,121905000,121900025 |
A0444363 | Certificate of Analysis | 20/07/2022 | 121900250,121900010,121901000,121905000,121900025 |
Safety and Handling

Product Identifier
- 8-Hydroxyquinoline
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction