Learn More
8-Methoxypsoralen, 99%, Thermo Scientific Chemicals
55.68 € - 180.90 €
Chemical Identifiers
CAS | 298-81-7 |
---|---|
Molecular Formula | C12H8O4 |
Molecular Weight (g/mol) | 216.19 |
MDL Number | MFCD00005009 |
InChI Key | QXKHYNVANLEOEG-UHFFFAOYSA-N |
Synonym | methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin |
PubChem CID | 4114 |
ChEBI | CHEBI:18358 |
IUPAC Name | 9-methoxyfuro[3,2-g]chromen-7-one |
SMILES | COC1=C2OC=CC2=CC2=C1OC(=O)C=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10368490
|
Thermo Scientific Acros
214150010 |
1 g | Glass bottle |
55.68 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10328540
|
Thermo Scientific Acros
214150050 |
5 g | Glass bottle |
180.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
298-81-7 | |
216.19 | |
QXKHYNVANLEOEG-UHFFFAOYSA-N | |
4114 | |
9-methoxyfuro[3,2-g]chromen-7-one |
C12H8O4 | |
MFCD00005009 | |
methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin | |
CHEBI:18358 | |
COC1=C2OC=CC2=CC2=C1OC(=O)C=C2 |
Specifications
298-81-7 | |
100.0 | |
White to Cream | |
98.5% min. (on anhydrous substance) (HPLC) | |
MFCD00005009 | |
15, 6059 | |
Solubility in water: practically insoluble in water. Other solubilities: sparingly soluble in boiling water,sparingly soluble in ether,soluble in boiling alcohol,acetone,acetic acid,,propylene glycol and benzene,freely soluble in chloroform | |
COC1=C2OC=CC2=CC2=C1OC(=O)C=C2 | |
216.19 | |
CHEBI:18358 | |
99% | |
8-Methoxypsoralen, 99% |
98.5 | |
143.0°C to 148.0°C | |
Authentic | |
C12H8O4 | |
19, I, 711 | |
methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin | |
QXKHYNVANLEOEG-UHFFFAOYSA-N | |
9-methoxyfuro[3,2-g]chromen-7-one | |
4114 | |
216.19 | |
Needle-like Crystals or Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-066-9
missing translation for 'rtecsNumber' : LV1400000
missing translation for 'tsca' : TSCA
- 8-Methoxypsoralen
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1A
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.