missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid
GREENER_CHOICE

Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.

Brand:  Thermo Scientific™ 8800

321.10 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11570115

  • 338.00 € / Pack of 20
Estimated Shipment: 05-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.

  • Molded with alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Natural
81 (1.5 or 2.0 mL Tubes)
Polypropylene
81-Well Cryogenic Storage Racks with Lid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid > Cryogenic Rack, Natural

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.