Learn More
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
17.00 € - 338.00 €
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
11570115
![]() |
Thermo Scientific™
8800 |
Natural |
338.00 €
Pack of 20 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10461411
![]() |
Thermo Scientific™
8810 |
Fluorescent Green |
17.00 €
Each |
Estimated Shipment: 14-10-2025
Log in to see stock. |
||||||
10781592
![]() |
Thermo Scientific™
8820 |
Fluorescent Orange |
338.00 €
Pack of 20 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
12358192
![]() |
Thermo Scientific™
8830-11 |
Fluorescent Pink |
17.30 €
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
||||||
Description
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
- Molded with alphanumeric grid for easy tube identification
- Lid protects samples and allows racks to be stacked
Specifications
Polypropylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid