Learn More
9-Bromo-1-nonene, 97%, Thermo Scientific Chemicals
42.81 € - 158.00 €
Chemical Identifiers
CAS | 89359-54-6 |
---|---|
Molecular Formula | C9H17Br |
Molecular Weight (g/mol) | 205.14 |
MDL Number | MFCD09037826 |
InChI Key | RQXPBVHYVAOUBY-UHFFFAOYSA-N |
Synonym | 9-bromo-1-nonene, 1-nonene, 9-bromo, 8-nonenyl bromide, 1-bromo-8-nonene, 9-bromanylnon-1-ene |
PubChem CID | 11019998 |
IUPAC Name | 9-bromonon-1-ene |
SMILES | BrCCCCCCCC=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15482149
|
Thermo Scientific Alfa Aesar
H64792.03 |
1 g |
42.81 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15492149
|
Thermo Scientific Alfa Aesar
H64792.06 |
5 g |
158.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
89359-54-6 | |
205.14 | |
RQXPBVHYVAOUBY-UHFFFAOYSA-N | |
11019998 | |
BrCCCCCCCC=C |
C9H17Br | |
MFCD09037826 | |
9-bromo-1-nonene, 1-nonene, 9-bromo, 8-nonenyl bromide, 1-bromo-8-nonene, 9-bromanylnon-1-ene | |
9-bromonon-1-ene |
Specifications
89359-54-6 | |
100°C to 101°C (15 mmHg) | |
Br(CH2)7CH=CH2 | |
MFCD09037826 | |
9-bromo-1-nonene, 1-nonene, 9-bromo, 8-nonenyl bromide, 1-bromo-8-nonene, 9-bromanylnon-1-ene | |
BrCCCCCCCC=C | |
205.14 | |
205.14 | |
Liquid |
Colorless to Yellow | |
C9H17Br | |
1.4675 | |
1746338 | |
RQXPBVHYVAOUBY-UHFFFAOYSA-N | |
9-bromonon-1-ene | |
11019998 | |
97% | |
9-Bromo-1-nonene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.