missing translation for 'onlineSavingsMsg'
Learn More
9-Fluorenylmethyl chloroformate, 98+%, Thermo Scientific Chemicals Product Code.: 11479173

9-Fluorenylmethyl chloroformate, 98+%, Thermo Scientific Chemicals

Product Code. 11479173
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11479173

Brand: Thermo Scientific Alfa Aesar A11683.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Store cold

Base sensitive amino protecting group for solid-phase peptide synthesis9-Fluorenylmethyl chloroformate is used in capillary electrophoresis. It acts as a reagent in the precolumn derivatization of amines for HPLC and fluorescent detection. Further, it is used for derivatizing amino acids for HPLC analysis. In addition to this, it is used to prepare N-Fmoc amino acids for solid-phase peptide synthesis and oligonucleotide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Base sensitive amino protecting group for solid-phase peptide synthesis9-Fluorenylmethyl chloroformate is used in capillary electrophoresis. It acts as a reagent in the precolumn derivatization of amines for HPLC and fluorescent detection. Further, it is used for derivatizing amino acids for HPLC analysis. In addition to this, it is used to prepare N-Fmoc amino acids for solid-phase peptide synthesis and oligonucleotide synthesis.

Solubility
Soluble in dioxane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28920-43-6
Molecular Formula C15H11ClO2
Molecular Weight (g/mol) 258.701
MDL Number MFCD00001138
InChI Key IRXSLJNXXZKURP-UHFFFAOYSA-N
Synonym 9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chlorideShow MoreShow Less
PubChem CID 34367
IUPAC Name 9H-fluoren-9-ylmethyl carbonochloridate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl

Specifications

Melting Point 60°C to 64°C
Assay Percent Range 98%
Quantity 5 g
UN Number UN2923
Beilstein 2279177
Sensitivity Moisture sensitive
Solubility Information Soluble in dioxane.
Formula Weight 258.7
Percent Purity 98%
Chemical Name or Material 9-Fluorenylmethyl Chloroformate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240445 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA11683.06,A11683.03,A11683.14
Lot Number10241707 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberA11683.06,A11683.03,A11683.14
Lot Number10231894 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberA11683.06,A11683.03,A11683.14
Lot Number10216189 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11683.06,A11683.03,A11683.14
Lot NumberF3508C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11683.06,A11683.03,A11683.14
compliance-icons compliance-icons
Product Identifier
  • 9-Fluorenylmethyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 9-Fluorenylmethyl chloroformate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9-Fluorenylmethyl chloroformate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.