Learn More
Fisherbrand™ 96 PP Well Plate with 96 Glass Insert and PE Snap Caps with Septum
For complex UHPLC, LC and GC applications
Brand:  Fisherbrand™ 08 20 0905
 View more versions of this product
Product Code. 12940851
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 96 Wells
  • Silicone/PTFE
  • Clear
  • Glass
  • Plates meet ANSI and SBS standards
  • Different sealing options
  • Chromatography tested
  • No leeching due to high-end material
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Clear
96
Round
8 x 12
Glass
1000 μL