missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural

Fit most thermal cyclers

109.00 € - 134.00 €

Specifications

Format Standard
Array 8 x 12
Product Type PCR Microplate
Skirt Style Skirted
Material Polypropylene
View More Specs

Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
12194112
View Documents
Fisherbrand™
14230241
Yellow
113.00 €
Pack of 25
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
 
12184112
View Documents
Fisherbrand™
14230240
Green
109.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
12164112
View Documents
Fisherbrand™
14230238
Red
113.00 €
Pack of 25
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
 
12154112
View Documents
Fisherbrand™
14230237
Natural
134.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
 
12174112
View Documents
Fisherbrand™
14230239
Blue
109.00 €
Pack of 25
Estimated Shipment: 05-08-2025
to see stock.
Add to basket
 
Description

Description

Fit most thermal cyclers

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Stackable
  • Raised-rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 200μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • SBS (Society for Biomolecular Screening) footprint

  • Automation friendly
  • Eight holes in the skirt aid plate positioning and removal from thermal cycler block
  • Cleanroom produced

Specifications

Specifications

Standard
PCR Microplate
Polypropylene
96
Yes
8 x 12
Skirted
200 μL
PCR, RT-PCR applications
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.