Learn More
Eppendorf™ 96-Well Protein LoBind Deep Well Plates
BIOPHARMAGREENER_CHOICE
Use these plates in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results. Eppendorf™ Deepwell Protein LoBind Plates recover significantly more protein for downstream analyses and enzymes remain active.
365.00 € - 795.00 €
Specifications
Format Standard
Array 8 x 12
No. of Wells 96
Dnase Free Yes
View More Specs
Products 2
Product Code Brand Volume (Metric) Well Color Packaging Price Quantity & Availability  
Product Code Brand Volume (Metric) Well Color Packaging Price Quantity & Availability  
10664473
BIOPHARMAGREENER_CHOICE
View Documents
Eppendorf™
0030504100
500 μL White 40 Plates (5 Bags x 8 Plates)
795.00 €
Pack of 40
In Stock
to see stock.
Edge
Add to basket
10115602
BIOPHARMAGREENER_CHOICE
View Documents
Eppendorf™
0030504216
1000 μL Yellow 20 Plates (5 Bags x 4 Plates)
365.00 €
Pack of 20
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
Description
Description
  • LoBind material guarantees maximum sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: of human DNA, DNAase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Unique OptiTrack™ matrix: 30% faster sample identification and fewer pipetting errors
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface guarantee reliable sealing in plates
Specifications
Specifications
Standard
8 x 12
96
Yes