missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates

Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.
Brand: Eppendorf™ 0030129563
1855.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- LoBind material guarantees maximum sample recovery for improved assay results.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).

Specifications
Includes Barcode | |
PCR Clean | |
Standard | |
8 x 12 | |
10 Bags x 30 Plates | |
Yellow | |
PCR Plate | |
Polypropylene, Polycarbonate | |
96 | |
Conical Bottom (V-well) | |
Yes | |
300 Pk. | |
PCR Clean (Non-sterile) | |
Yes |
Low DNA Binding | |
Yes | |
With Lid | |
Autoclavable | |
Eppendorf | |
Single-use | |
Skirted | |
150 μL | |
Non-sterile | |
Real Time PCR | |
6000 x g | |
Yes | |
300 Pk. | |
Colorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates > No. of Wells: 96, Well Volume: 150 μL, Skirt Style: Skirted, Color: Yellow, Packaging: 300 Plates (10 Bags x 30 Plates)
Spot an opportunity for improvement?Share a Content Correction