missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates

Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.
192.77 € - 1955.20 €
Specifications
Format | Standard |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene, Polycarbonate |
Sterility | Non-sterile |
Purity or Quality Grade | PCR Clean (Non-sterile) |
Product Code | Brand | Color | No. of Wells | Volume (Metric) Well | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | No. of Wells | Volume (Metric) Well | Packaging | Price | Quantity & Availability | |||
15280735
![]() |
Eppendorf™
0030129512 |
Clear | 96 | 150 μL | 5 Bags x 5 Plates |
192.77 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||
15891525
![]() |
Eppendorf™
0030129580 |
Blue | 96 | 150 μL | 5 Bags x 5 Plates |
202.00 €
Pack of 25 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||
15801535
![]() |
Eppendorf™
0030129598 |
Red | 96 | 150 μL | 5 Bags x 5 Plates |
202.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
15821535
![]() |
Eppendorf™
0030129660 |
Green | 96 | 150 μL | 5 Bags x 5 Plates |
202.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
15831535
![]() |
Eppendorf™
0030129679 |
Yellow | 96 | 150 μL | 5 Bags x 5 Plates |
202.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
15861525
![]() |
Eppendorf™
0030129555 |
Green | 96 | 150 μL | 10 Bags x 30 Plates |
1880.00 €
Pack of 300 |
In Stock
Log in to see stock. |
|||
15871525
![]() |
Eppendorf™
0030129563 |
Yellow | 96 | 150 μL | 10 Bags x 30 Plates |
1955.20 €
Pack of 300 |
In Stock
Log in to see stock. |
|||
15881525
![]() |
Eppendorf™
0030129571 |
Orange | 96 | 150 μL | 300 Plates |
1880.00 €
Pack of 300 |
In Stock
Log in to see stock. |
|||
15277803
![]() |
Eppendorf™
0030129504 |
Clear | 96 | 250 μL | 5 Bags x 5 Plates |
192.77 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||
15911992
![]() |
Eppendorf™
0030129547 |
Clear | 384 | 45 μL | 5 Bags x 5 Plates |
285.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
Description
- LoBind material guarantees maximum sample recovery for improved assay results.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
Specifications
Standard | |
Polypropylene, Polycarbonate | |
PCR Clean (Non-sterile) |
Autoclavable | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates