Learn More
Uvex™ 9906 Pheos Polycarbonate Visors
Visors are treated with supravision excellence coating for anti-fog and anti-scratch protection. Available with two alternative mountings: mechanical levers or automatic magnetic hooks.
Brand:  Uvex™ 9906002
47.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17250272
  • 55.45 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Magnetic locking system is extremely easy to attach to the visor
  • Ergonomic joint system is comfortable to wear, even for long periods
  • Compatible with Pheos B-S-WR and Pheos Alpine (Adapter 9790067 is required for combination with Alpine)
  • Visor 9906002 can be combined with earmuffs Pheos K2H Earmuffs (Mechanical)
  • Visor 9906003 is compatible with Pheos K2H Earmuffs (Magnetic)
  • Certified for areas of application 3 (liquids) and 9 (molten metal/hot solids)
  • Labeling: W 166 3 8-2 9 B CE 0196 (Frame), 2C-1.2 W 1 B 9 KN CE 0196 (Lens)
  • Lens can be changed
  • Helmet sold separately
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 166:2001, EN 170:2002
Pheos B-S-WR, Pheos Alpine (Requires Adaptor 9790067), and Pheos K2H Earmuffs (Mechanical)
Polycarbonate
Mechanical Levers
Black
Clear
9906 Pheos
Visor