Learn More
Fisherbrand™ 9mm Short Thread Seal, Transparent, MS-Cap, thinned penetration area
Polypropylene caps are chemically inert and suitable for most chromatography applications.
Brand:  Fisherbrand™ 09 08 2000
Product Code. 11894820
  • 367.00 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Elimination of septum push-through
  • Increased sealing capability
  • Improved autosampler compatibility
  • Flexibility to select the best septum for your instrument and applications
  • Optimized ergonomics, fine texturing and evenly spaced ribbing for superior handling
  • Easy-on, easy-off convenience with just one turn
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Closures have the profile of a crimp or snap closure for compatibility with robotic autosamplers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Short Thread Cap
100
Screw Cap
9 mm
GC, HPLC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ 9mm Short Thread Seal, Transparent, MS-Cap, thinned penetration area >