Learn More
Labconco™ Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Holds tubes in upright position for freeze drying.
Brand:  Labconco™ 7442700
 View more versions of this product
Product Code. 10316372
  • 341.00 € / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Holds tubes in upright position for freeze drying.
Hold tubes in upright position inside 900mL or larger Fast-Freeze Flasks and 750mL or larger Lyph-Lock Flasks Stainless steel
Specifications
Specifications
Tube Holder
Stainless Steel
900ml or larger fast freeze™ flasks and 750ml or larger lyph lock flasks
8.33 cm
13.9 cm
0.5 kg
750, 900 mL
Hold tubes in upright position
3.5 in.
5 in.
0.5 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks > Holds 2mL tubes