missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AbC™ Total Antibody Compensation Bead Kit

Provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated mouse, rat, hamster, or rabbit antibodies with any laser in flow cytometry.

Brand:  Invitrogen™ A10513

132.05 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 15308666

  • 139.00 € / 25 tests

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Broadest multispecies reactivity—for use with all isotypes of mouse, rat, hamster, and rabbit antibody conjugates
  • Brightest signal—most sensitive antibody-capture beads available
  • Maximum laser compatibility—for use with all lasers
  • Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads).
  • Ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3), rat (IgG1, IgG2a, IgG2b), hamster (Armenian IgG and Syrian IgG), and rabbit (monoclonal and polyclonal) immunoglobulins.Demonstrates the highest and most consistent reactivity to the different subclasses of immunoglobulin in the industry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • 6 μm
    Flow Cytometer
    25 Tests
    Room Temperature
    Compensation Beads
    Fluorescence
    any lasers (UV to 633/635)
    Tube
    25 tests
    AbC™
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.