missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ABsolute QPCR Mix, ROX
Optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes.
596.00 € - 2205.00 €
Specifications
Detection Method | Primer-probe |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11847682
|
Thermo Scientific™
AB1138B |
1600 Reactions |
2205.00 €
1600 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11857682
|
Thermo Scientific™
AB1139A |
400 Reactions |
596.00 €
400 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.
- High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
- Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
- Optimized buffer - For maximal performance in multiplex reactions
Specifications
Primer-probe | |
qPCR | |
Standard | |
ABsolute | |
Gene Expression | |
2 X |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System | |
qPCR Mix | |
2X | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title