missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute QPCR Mix, SYBR Green, low ROX

Optimized for SYBR Green chemistry and contains all the components necessary to perform quantitative PCR, with the exception of template and primers.

Brand:  Thermo Scientific™ AB1323A

350.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11802093

  • 700.00 € / 400 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.

ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.

  • High Sensitivity & Specificity - The master mixes contain the patented hot start enzyme Thermo-Start DNA Polymerase
  • Optimized buffer containing SYBR Green
  • Reproducibility - All master mixes undergo extensive QC to ensure batch to batch consistency
  • Broad dynamic range
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 1 x 5 mL ABsolute QPCR SYBR Green Low ROX Mix (2X)
• 100 μL MgCl2 (1M)

Store at -20°C
qPCR
Standard
ABsolute
400 reactions
Gene Expression
2X
DNA, dsDNA
SYBR
Taq DNA Polymerase
7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
Real Time PCR SYBR Master Mix
400 Reactions of 25 μL Each
qPCR Cyclers
400 Reactions
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.