Learn More
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers
Get maximum flexibility, regardless of your application, by using these accessories for Thermo Scientific™ MaxQ™ High Performance orbital shakers.
Brand:  Thermo Scientific™ 452000
 View more versions of this product
Product Code. 10474432
  • 4864.20 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shaker Kit with Mount and Stand
Dual-stack benchtop shaker kit; Includes kneewell side mount and orbital shaker stand
MaxQ™ HP 420 Shaker