Learn More
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers
Get maximum flexibility, regardless of your application, by using these accessories for Thermo Scientific™ MaxQ™ High Performance orbital shakers.
19.90 € - 4864.20 €
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
12602795
|
Thermo Scientific™
194040 |
Quick change platform adapter | MaxQ HP 416, 430, 435, 436, 480, 481 |
1015.05 €
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||
12652785
|
Thermo Scientific™
194045 |
Gassing manifold (one gas) | MaxQ HP 420, 435, 436, 480, 481 |
681.39 €
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||
12632795
|
Thermo Scientific™
194139 |
Magnote wipe-off magnet and pen kit (floor models) | MaxQ™ HP 435, 436, 480, 481 Shaker |
19.90 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
12612795
|
Thermo Scientific™
194140 |
Magnote wipe-off magnet and pen kit (benchtop models) | MaxQ™ HP 416, 430, 420 Shaker |
21.81 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
10474432
|
Thermo Scientific™
452000 |
Dual-stack benchtop shaker kit; Includes kneewell side mount and orbital shaker stand | MaxQ™ HP 420 Shaker |
4864.20 €
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||
10719273
|
Thermo Scientific™
201127 |
7 Day Recorder, 220V | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |
2361.75 €
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||
13475989
|
Thermo Scientific™
760440 |
Replacement HEPA filter | MaxQ HP 435, 436, 480, 481, 8000 |
218.09 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||
Description
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.