Learn More
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Generate an accurate turbidity reading quickly with this reliable Turbidity Meter and its accessories.
Brand:  Thermo Scientific™ AC3CBR
 View more versions of this product
Product Code. 12043273
  • 31.26 € / Pack of 10
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Features:
  • Accessories for AQUAfast AQ3010 Turbidity Meter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AQUAfast AQ3010 Turbidity Meter
Vial Cleaning Brushes, 5 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter >