Learn More
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Generate an accurate turbidity reading quickly with this reliable Turbidity Meter and its accessories.
31.26 € - 81.61 €
Specifications
For Use With (Equipment) AQUAfast AQ3010 Turbidity Meter
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12043273
View Documents
Thermo Scientific™
AC3CBR
Vial Cleaning Brushes, 5 Pack
31.26 €
Pack of 10
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
12033273
View Documents
Thermo Scientific™
AC3SIL
Silicone Oil (1mL Bottle) And Cloth
42.81 €
Each
12023273
View Documents
Thermo Scientific™
AC3V25
25mm Vials, 3 Pack
81.61 €
Pack of 3
In Stock
to see stock.
Add to basket
Description
Description
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Additional Accessory: 
  • Replacement Calibration Standards (see AC301S)
Specifications
Specifications
AQUAfast AQ3010 Turbidity Meter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter